Test tube holders, wood
Supplier: GLASWARENFABRIK KARL HECHT
42811010
HECH42811010EA 3.1 SGD
HECH42811010
Test tube holders, wood
Tongs
Wooden, with spring.
  • Peg/clip shape
Suitable for tubes up to 20/30 mm diameter.