Instrument trays with lid
Supplier: GLASWARENFABRIK KARL HECHT
42915003
HECH42915003EA 118 SGD
HECH42915003
Instrument trays with lid
Trays/Dishes/Pots Instrument Trays/Dishes
Made of stainless steel.
  • Resistant to hot-air sterilization up to +180 °C
  • Excellent chemical resistance
  • With knob lid