Absorption Cells with Lids
Supplier: HELLMA
104-10-40
HELL104-10-40EA 544 SGD
HELL104-10-40
Absorption Cells with Lids
Cuvettes Spectrophotometer Cuvettes
Hellma® high precision quartz glass semi-micro cells for absorption measurements in the UV/Vis range, 10 to 50 mm path length, with lid.
  • Cells with PTFE lid
  • 10 to 50 mm path length
  • Highest precision
  • Excellent transmission
  • Sustainably produced
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
Highest precision: Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement results.