Accessories for 1300 Series Class II, Type A2 Biological Safety Cabinets
Supplier: Thermo Scientific
1911322
HERA1911322EA 0 SGD
HERA1911322
Accessories for 1300 Series Class II, Type A2 Biological Safety Cabinets
Fume Hoods and Enclosures Biological Safety Cabinets
Stands - Rolling caster stand provides comfortable fixed worksurface height of 34 inches (86 cm) for seated applications; designed for limited cabinet mobility for cleaning purposes.
Certifications: NSF/ANSI 49, Class II Type A2.
Order Now
SPECIFICATIONS
Environmentally Preferable