CO₂ incubators, BBD 6220 and Cytoperm® 2
Supplier: Thermo Scientific
51011659 51011660 51020241
HERA51011659EA 0 SGD
HERA51011659 HERA51011660 HERA51020241
CO₂ incubators, BBD 6220 and Cytoperm® 2
Incubators CO₂ Incubators
These CO₂ incubators embody an innovative generation of equipment offering optimum protection from contaminants. They guarantee optimum repeatability of results. Thanks to their outstanding performance features the BBD 6220 and the Cytoperm®2 are predestined for all sectors making stringent demands on protection, safety and quality of work: in microbiological laboratories and with in vitro fertilisation as well as in cancer research or tissue engineering. Each incubator is characterised by reliability, safety and practical application characteristics complying with highest levels of quality.
More Product Information