Heracell™ VIOS CO₂ incubators
Supplier: Thermo Scientific
51900293 51901126 51901144 51033549 51033559
HERA51900293EA 0 SGD
HERA51900293 HERA51901126 HERA51901144 HERA51033549 HERA51033559
Heracell™ VIOS CO₂ incubators
Incubators CO₂ Incubators
The Heracell™ VIOS series has been designed for sensitive cultures like stem and primary cells in leading research, pharmaceutical and clinical applications. The units are available with either electropolished stainless steel or 100% pure copper interiors and have adjustable, perforated shelving, easy to clean corners with convenient access port and a reversible exterior door for added flexibility.
  • THRIVE active airflow maintains homogeneous conditions avoiding unwanted sample variation with fast recovery times of under 10 minutes utilising long life,
More Product Information