Megafuge® 8/8R Centrifuges, Bench Top, Ventilated/Refrigerated
Supplier: Thermo Scientific
75007211
HERA75007211EA 0 SGD
HERA75007211
Megafuge® 8/8R Centrifuges, Bench Top, Ventilated/Refrigerated
Centrifuges Benchtop Centrifuges
These durable centrifuges are available as ventilated or refrigerated versions and provide exceptional capacity in a compact design with a smart, simple interface.
  • Quiet performance provides a safe, stress-free environment
  • One touch operation with pre-saved protocols and password protection
  • Highly visible backlit display for easy reading of parameters
  • Secure, push-button Auto-Lock™ function enables quick and easy rotor exchange and clear chamber access for cleaning convenience
  • Biocontainment sealing options, including certified ClickSeal® lids for glove
More Product Information