Storage Racks for Ultra-Low Temperature Freezers
Supplier: Thermo Scientific
920090
HERA920090EA 0 SGD
HERA920090
Storage Racks for Ultra-Low Temperature Freezers
Racks Cryogenic Box Racks
Cryobox storage racks makes organising your ultra-low temperature freezer samples easy.
  • Sliding drawer racks
  • Adjustable side access box racks
  • Stores 5 cm (2") and 7.6 cm (3") cryo boxes and microplates
Certifications:
cULus.
Order Now
SPECIFICATIONS
Environmentally Preferable