VWR® Accessory Shelf Kits for Ultra-Low Temperature Freezers, Avantor
Supplier: Thermo Scientific
SK600TSX
HERASK600TSXEA 0 SGD
HERASK600TSX
VWR® Accessory Shelf Kits for Ultra-Low Temperature Freezers, Avantor
Freezers Ultra Low Temperature Freezers
Organize your ULT freezer storage with additional interior stainless steel shelves.
  • Stainless steel shelf kit by freezer size
  • Includes shelf clips for installation
Certifications:
cULus and CE.
Packaging:
One ULT shelf and installation clips provided.