High Performance Lab Refrigerators, TSX
Supplier: Thermo Scientific
TSX2305GV
HERATSX2305GVEA 12660 SGD
HERATSX2305GV
High Performance Lab Refrigerators, TSX
Refrigerators
Available in glass or solid door, the TSX series high-performance refrigerators combine advanced microprocessor temperature control with high-quality cabinet construction and alarm systems. Positive, forced-air circulation is designed to maintain temperature uniformity to protect important medical and pharmaceuticalgrade storage as well as laboratory media, reagents and more. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.
  • Forced-air circulation
More Product Information