Ultra-Low Temperature Freezers, TSX
Supplier: Thermo Scientific
TSX60086V
HERATSX60086VEA 0 SGD
HERATSX60086V
Ultra-Low Temperature Freezers, TSX
Freezers Ultra Low Temperature Freezers
Thermo Scientific™ TSX series ultra-low high performance freezers are designed with features that support sustainability objectives without compromising performance. The series' V-drive technology is designed to provide temperature uniformity that adapts to the laboratory environment, offering significant energy savings. Now available in four capacities, these −80 °C freezers maximise sample storage capacity and offer substantial energy savings over conventional refrigerant ultra-low freezers.
  • Best peak variation of energy efficient, ultra-low freezers: Less than 5 °C
  • Record
More Product Information