Vydac 202TP C18-HL, HPLC columns
Supplier: Avantor
HICH202TP54EA 1980 SGD
HICH202TP54
Vydac 202TP C18-HL, HPLC columns
Chromatography Columns
Vydac TP is the first range of wide pore media phases developed in the Vydac range and it is the industry's standard for peptide and protein separations. The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L1
Higher carbon load than 201TP. Developed for the separation of PAHs.
* Use with holder 80More Product Information