Avantor® Hichrom Carbohydrate Cation, HPLC Column
Supplier: Avantor
HICH70057EA 6750 SGD
HICH70057
Avantor® Hichrom Carbohydrate Cation, HPLC Column
Chromatography Columns
This column consists of a highly efficient sulphonated polystyrene resin supplied in the calcium form. It provides excellent separations using only water as the mobile phase.
  • Sulphonated polystyrene resin in the calcium form
  • 100% water used as mobile phase
  • Column heating required
  • USP L19