Avantor® Alltima HP, HPLC columns, C18-Amide, 3 µm
Supplier: Avantor
HICH87607EA 1690 SGD
HICH87607
Avantor® Alltima HP, HPLC columns, C18-Amide, 3 µm
Chromatography Columns
Avantor® Alltima HP columns has polar-embedded phase with extremely low bleed.
  • High resolution for complex samples
  • High purity silica
  • Excellent column stability
These columns combine the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
These are widely used for basic compounds in neutral to alkaline pH, MS applications.