Avantor® Alltima HP, HPLC Columns, C18-HL, 3 µm
Supplier: Avantor
HICH22194EA 0 SGD
HICH22194 HICH22195 HICH87506 HICH87688 HICH87610 HICH87827 HICH87686 HICH87690 HICHAHP-3C18H-250D
Avantor® Alltima HP, HPLC Columns, C18-HL, 3 µm
Chromatography Columns
Avantor® Alltima HP columns contain the highest carbon load for superior retention and loadability.
  • High resolution for complex samples
  • High purity silica
  • Excellent column stability
  • Low to no detectable column bleed
These columns combine the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.