Avantor® Prevail Silica, HPLC Columns
Supplier: Avantor
HICH99341EA 0 SGD
HICH99341
Avantor® Prevail Silica, HPLC Columns
Chromatography Columns
Prevail™ HPLC columns exhibit long column lifetime in both highly aqueous and highly organic mobile phases. All phases, apart from Prevail™ Carbohydrate ES are silica-based. The stability of these phases is such that a single column can be switched between highly aqueous for analysis of highly polar analytes and highly organic for strong retention of hydrophobic analytes.
  • Highly polar phase
  • Stable from highly organic to highly aqueous
  • Speciality phases for specific applications
  • Excellent sensitivity with microbore and ELSD applications
These columnsMore Product Information