Avantor® Genesis, HPLC Columns, Silica
Supplier: Avantor
HICHFM25961EEA 0 SGD
HICHFM25961E
Avantor® Genesis, HPLC Columns, Silica
Chromatography Columns
Avantor® Genesis phases are based on high purity metal-free spherical silica. They are suitable for the analysis of a wide range of analytes.
  • Excellent peak shape and efficiency
  • Long column life
Order Now
SPECIFICATIONS
Environmentally Preferable
Surface area 300 m²/g
USP code L3