Avantor® Hichrom PEEKsil™, Tubing Sleeves, PEEK, MicroTight
Supplier: Avantor
HICHHI-171EA 145 SGD
HICHHI-171
Avantor® Hichrom PEEKsil™, Tubing Sleeves, PEEK, MicroTight
Tubing Chromatography Tubing
PEEK.
  • Colour-coded for easy Int.Ø identification
  • Temperature stable up to 125 °C
These tubing sleeves are used with MicroTight fittings to connect capillary tubing. The Int.Ø of the sleeve should be 0,001 to 0,002" larger than the Ext.Ø of the capillary tubing.
Order Now
SPECIFICATIONS
Environmentally Preferable