VWR® Hichrom Pliers, for Stainless Steel Tubing
Supplier: Avantor
HICHHI-196EA 205 SGD
HICHHI-196
VWR® Hichrom Pliers, for Stainless Steel Tubing
Pliers
These pliers are designed for cutting 1/16" stainless steel tubing and are especially suitable for tubing of 0,010" and 0,020" ID.
  • Leaves smooth and uniform cuts
  • De-burring may be necessary to achieve an optimum finish