Avantor® Hichrom, Frit Removal Tool
Supplier: Avantor
HICHHI-300EA 386 SGD
HICHHI-300
Avantor® Hichrom, Frit Removal Tool
Filters Precolumn Filters and Frits
The frit removal tool is ideal for the removal of frits used in HPLC/LC columns.