Avantor® Partisil ODS, HPLC Columns
Supplier: Avantor
HICHP10ODS-200AEA 1720 SGD
HICHP10ODS-200A HICHP10ODS-250AF HICHP10ODS-50A HICHP10ODS-10C5-SK HICHP10ODS-300AF
Avantor® Partisil ODS, HPLC Columns
Chromatography Columns
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity for preparative work.
  • Irregular porous silica
  • Convenient and easy to use: Hand tightened fittings
  • Increased column lifetime
Low carbon load C18 phase, with intentionally unbonded surface silanols adding to the selectivity.
Caution: *Use with freestanding holder HI-161 and column coupler HI-081, please see "HPLC Guard Column Holders and Accessories"