Avantor® Partisil ODS3, HPLC Columns
Supplier: Avantor
HICHP10ODS3-250AEA 1076 SGD
HICHP10ODS3-250A HICHP5ODS3-150A HICHP5ODS3-250A HICHP10ODS3-10C5 HICHP5ODS3-10C5 HICHP10ODS3-150A HICHP5ODS3-100A HICHP5ODS3-250AF HICHP5ODS3-125A HICHP10ODS3-300AF
Avantor® Partisil ODS3, HPLC Columns
Chromatography Columns
Avantor® Partisil® was one of the first commercially available irregular silicas with a large surface area giving it a high loading capacity.
  • Easy to use
  • Irregular porous silica
Avantor® Partisil® ODS3 is an intermediate carbon load C18 phase used for pharmaceutical, natural products, food, biological and environmental pollutants applications.