Polygosil 60-5 C18, HPLC Columns
Supplier: Avantor
HICHPOLY-5C18-150AEA 0 SGD
HICHPOLY-5C18-150A
Polygosil 60-5 C18, HPLC Columns
Chromatography Columns
Irregular silica phases for analytical applications.
  • pH stability: 2 to 8