Demeter pipettes
Supplier: HIRSCHMANN
1500137
HIRS1500137EA 27.6 SGD
HIRS1500137
Demeter pipettes
Pipettes Graduated Pipettes
AR-Glas®.
  • Calibrated to deliver 'Ex'
  • Blue graduation
  • Dilution pipettes with two marks at one and one,1 ml or with four marks at 1,0/2,0/2,1/2,2 ml
Length: 250 mm ±5 mm
Length: 250 mm ±5 mm
Certifications: DIN 12838 and DIN 12839