Measuring cylinders, tall form, class B, borosilicate glass
Supplier: HIRSCHMANN
2230194 2230154 2230160 2230170 2230175 2230180 2230186 2230190 2230193
HIRS2230194EA 230 SGD
HIRS2230194 HIRS2230154 HIRS2230160 HIRS2230170 HIRS2230175 HIRS2230180 HIRS2230186 HIRS2230190 HIRS2230193
Measuring cylinders, tall form, class B, borosilicate glass
Cylinders Measuring Cylinders
DURAN®, borosilicate glass, tall form, class B.
  • Calibrated to contain 'ln'
  • Hexagonal glass base, with spout
  • Short graduation marks, blue graduations
DIN EN ISO 4788
Certifications: DIN EN ISO 4788
Order Now
SPECIFICATIONS
Environmentally Preferable