Measuring cylinders, low form, class B, borosilicate glass
Supplier: HIRSCHMANN
2400194 2400160 2400170 2400175 2400180 2400186 2400190 2400193
HIRS2400194EA 212 SGD
HIRS2400194 HIRS2400160 HIRS2400170 HIRS2400175 HIRS2400180 HIRS2400186 HIRS2400190 HIRS2400193
Measuring cylinders, low form, class B, borosilicate glass
Cylinders Measuring Cylinders
DURAN®, borosilicate glass, low form, class B.
  • Calibrated to contain 'In'
  • Hexagonal base with spout
  • Marks and inscriptions in amber stain, short graduation marks
DIN EN ISO 4788
Order Now
SPECIFICATIONS
Environmentally Preferable