Volumetric flasks, class A, white markings
Supplier: HIRSCHMANN
2640152 2640153 2640160 2640161 2640165 2640170 2640175 2640178 2640180 2640181 2640185 2640186 2640190 2640193 2640194
HIRS2640152EA 113 SGD
HIRS2640152 HIRS2640153 HIRS2640160 HIRS2640161 HIRS2640165 HIRS2640170 HIRS2640175 HIRS2640178 HIRS2640180 HIRS2640181 HIRS2640185 HIRS2640186 HIRS2640190 HIRS2640193 HIRS2640194
Volumetric flasks, class A, white markings
Flasks Volumetric Flasks
DURAN®, borosilicate glass, amber, with PE stopper, conformity certified (DE-M marking).
  • White enamel indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck.
Certifications: DIN EN ISO 1042
Order Now
SPECIFICATIONS
Environmentally Preferable