Volumetric flasks, class A, amber markings
Supplier: HIRSCHMANN
2822166 2822153 2822161 2822165 2822170 2822175 2822178 2822180 2822181 2822185 2822186 2822190 2822193 2822194 2822160 2822171
HIRS2822166EA 28.8 SGD
HIRS2822166 HIRS2822153 HIRS2822161 HIRS2822165 HIRS2822170 HIRS2822175 HIRS2822178 HIRS2822180 HIRS2822181 HIRS2822185 HIRS2822186 HIRS2822190 HIRS2822193 HIRS2822194 HIRS2822160 HIRS2822171
Volumetric flasks, class A, amber markings
Flasks Volumetric Flasks
DURAN®, borosilicate glass, with PE stopper, conformity certified (DE-M marking).
  • Amber stain markings
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck
Certifications: DIN EN ISO 1042