Volumetric flasks, class A, blue markings
Supplier: HIRSCHMANN
2950186 2950190
HIRS2950186EA 552 SGD
HIRS2950186 HIRS2950190
Volumetric flasks, class A, blue markings
Flasks Volumetric Flasks
DURAN®, borosilicate glass, with rim.
  • Blue enamelled indelible markings resistant to acids and alkalis
  • Conformity certified (DE-M marking) with dated batch identification
  • In accordance with the prepacking regulations
Order Now
SPECIFICATIONS
Environmentally Preferable