Silicone stoppers
Supplier: HIRSCHMANN
8905765 8905928
HIRS8905765EA 835 SGD
HIRS8905765 HIRS8905928
Silicone stoppers
Stoppers Silicone Stoppers
SILICOSEN® and BIO-SILICO® sterile stoppers are manufactured in a special process and have regular pores, thus making them ideal for the preparation and sterilisation of culture media. The uniform pores and good permeability to air make the stoppers ideally suitable for cultivation of aerobic microorganisms.
  • Chemical and temperature resistant
  • Liquid repellent
  • Reusable
SILICOSEN® T-type and L-type: Low humidity evaporation reduces dehydration of cultures.
SILICOSEN® C-type: Excellent permeability and secure fitting, thanks to seal lip. Ideally suitableMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable