Tubing for rotarus® peristaltic pumps, Tygon® F-4040-A
Supplier: HIRSCHMANN
95219016 95219015 95219013 95219017 95219503 95219725 95219717 95219710 95219703 95219522 95219517 95219510
HIRS95219016EA 812 SGD
HIRS95219016 HIRS95219015 HIRS95219013 HIRS95219017 HIRS95219503 HIRS95219725 HIRS95219717 HIRS95219710 HIRS95219703 HIRS95219522 HIRS95219517 HIRS95219510
Tubing for rotarus® peristaltic pumps, Tygon® F-4040-A
Tubing Transfer Tubing
These tubes are used for hydrocarbons and mineral oil products due to its low gas permeability, ozone resistant.
  • Temperature resistance from –40 to 75 °C
  • Hardness (Shore durometer A): 57