Tubing for rotarus® peristaltic pumps, Tygon® LFL
Supplier: HIRSCHMANN
95223011
HIRS95223011EA 1580 SGD
HIRS95223011
Tubing for rotarus® peristaltic pumps, Tygon® LFL
Tubing Transfer Tubing
Long flex life pump tube for general applications in the lab and industry and a long service life with maximum bending fatigue strength of all transparent Tygon® tubes, extremely low particle detachment, broad chemical resistance, long service life.
  • Temperature resistance from –54 to 74 °C
  • Hardness (Shore durometer A): 56
Order Now
SPECIFICATIONS
Environmentally Preferable