Ammonium formate ≥97.0% (by titrimetric analysis), Purum p.a.

Supplier: Honeywell Chemicals
Warning

Synonyms: Formic acid ammonium salt

09739-100G
HONC09739-100GEA 183 SGD
HONC09739-100G
Ammonium formate ≥97.0% (by titrimetric analysis), Purum p.a.
Ammonium formate

  • Products with a guarantee certificate and/or suitable for the stated analytical application.

Formula: HCO₂NH₄
MW: 63,06 g/mol
Melting Pt: 116 °C
Density: 1,266 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00013103
CAS Number: 540-69-2
EINECS: 208-753-9
Merck Index: 13,00525

Order Now

Specification Test Results

Identity (IR) complying
Assay 97 - 103 %
pH (5 %, 25 °C) 6.0 - 7.0
Colour of the solution complying
Turbidity of solution complying
Water (Karl Fischer) Max. 3 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 50 ppm
Cobalt (Co) Max. 50 ppm
Copper (Cu) Max. 50 ppm
Iron (Fe) Max. 50 ppm
Potassium (K) Max. 200 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 50 ppm
Zinc (Zn) Max. 50 ppm
Chloride (Cl) Max. 50 ppm
Sulfate (SO₄) Max. 100 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR