Iron(III) chloride 45% (FeCl₃ basis) in aqueous solution, Purum, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Iron trichloride, Ferric chloride

12322-4X2.5L
HONC12322-4X2.5LEA 3020 SGD
HONC12322-4X2.5L
Iron(III) chloride 45% (FeCl₃ basis) in aqueous solution, Purum, Fluka™
Iron (III) chloride

  • ≥95% for general laboratory use.

Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 315
Melting Pt: 260
Density: 1.48 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8 III
Merck Index: 13,04048

Order Now

Specification Test Results

Assay (excluding Fe) 44.6 - 45.4 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR