Aluminium potassium sulphate dodecahydrate 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., Fluka™

Supplier: Honeywell Chemicals

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium

12623-1KG
HONC12623-1KGEA 416 SGD
HONC12623-1KG
Aluminium potassium sulphate dodecahydrate 99.0-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., Fluka™
Aluminium potassium sulphate dodecahydrate
Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Melting Pt: 92.5 °C
Density: 1.75 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149143
CAS Number: 7784-24-9
EINECS: 233-141-3
Merck Index: 12,00373

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.0 - 100.5 %
Loss on drying 43.0 - 46.0 %
Ammonium (NH₄) Max. 0.2 %
Iron (Fe) complying
Arsenic (As) Max. 0.0002 %
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 0.005 %
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR