Potassium sulphate 99-101, meetSpec_Ph.Eur., Fluka™

Supplier: Honeywell Chemicals

12658-50KG 12658-1KG
HONC12658-50KGEA 4900 SGD
HONC12658-50KG HONC12658-1KG
Potassium sulphate 99-101, meetSpec_Ph.Eur., Fluka™
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1.698 °C (1.013 hPa)
Melting Pt: 1.067 °C
Density: 2.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 13,07759

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.0 - 101.0 %
Loss on drying (130°C) Max. 0.5 %
Acidity or alkalinity complying
Calcium (Ca) Max. 50 ppm
Iron (Fe) Max. 10 ppm
Magnesium (Mg) Max. 20 ppm
Sodium (Na) Max. 100 ppm
Heavy metals (as Pb) Max. 0.001 %
Chloride (Cl) Max. 20 ppm
Appearance of the solution complying
Residual solvents complying

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR