Benzyl alcohol ≥99.0% (by GC), Puriss. p.a. ACS analytical reagent, Riedel-de Haën™

Supplier: Honeywell Chemicals
Warning

Synonyms: Benzenemethanol, Phenylmethanol, Phenylcarbinol

13160-2.5L 13160-500ML
HONC13160-2.5LEA 1620 SGD
HONC13160-2.5L HONC13160-500ML
Benzyl alcohol ≥99.0% (by GC), Puriss. p.a. ACS analytical reagent, Riedel-de Haën™
Benzyl alcohol

ACS GENERAL USE SOLVENTS


  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested to meet ACS general use specifications


Beilstein Number: 878307
RTECS Number: DN3150000
UNSPSC Code: 12191503

Formula: C₆H₅CH₂OH
Boiling Pt: 205,3 °C (1.013 hPa)
Melting Pt: -15,4 °C
Density: 1,045 g/cm³ (20 °C)
Flash Pt: 100,4 °C
Storage Temperature: Ambient
MDL Number: MFCD00004599
CAS Number: 100-51-6
EINECS: 202-859-9
Merck Index: 13,01126
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0c85c958ac64f8b86c5836e49f2c4354b22886d&vid=633ad635461465914e782d5a22480c9ff8f81bf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.540 - 1.542
Non-volatile matter Max. 0.005 %
Sulfated ash Max. 0.005 %
Acetophenone (GC) Max. 0.02 %
Benzaldehyde (GC) Max. 0.01 %
Free acid (as C₆H₅COOH) Max. 0.03 %
Appearance of the solution complying
Aluminium (Al) Max. 0.5 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.5 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 2 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.1 ppm
APHA Max. 20


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR