Potassium permanganate 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™

Supplier: Honeywell Chemicals
Danger

13206-1KG 13206-25KG 13206-2.5KG 13206-250G
HONC13206-1KGEA 519 SGD
HONC13206-1KG HONC13206-25KG HONC13206-2.5KG HONC13206-250G
Potassium permanganate 99-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., USP, Fluka™
Potassium permanganate
Formula: KMnO₄
Melting Pt: Dec.
Density: 2,700 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1,II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay 99.0 - 100.5 %
Assay (calculated to the dried substance) 99.0 - 100.5 %
Water insoluble matter Max. 0.2 %
Loss on drying (on silica gel, 18h) Max. 0.2 %
Chloride (Cl) Max. 100 ppm
Sulfate (SO₄) Max. 500 ppm
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR