Sodium carbonate, anhydrous 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., NF, FCC, E500, Fluka™

Supplier: Honeywell Chemicals
Warning

13418-5KG 13418-25KG 13418-1KG
HONC13418-5KGEA 1170 SGD
HONC13418-5KG HONC13418-25KG HONC13418-1KG
Sodium carbonate, anhydrous 99.5-100.5% (dried basis), Puriss., meets analytical specification of BP, Ph. Eur., NF, FCC, E500, Fluka™
Sodium carbonate
Formula: Na₂CO₃
Melting Pt: 854 °C
Density: 2,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (calculated to the dried substance) 99.5 - 100.5 %
Loss on drying (300°C) Max. 0.5 %
Loss on drying (105°C, 4 h) Max. 0.5 %
Arsenic (As) Max. 2 ppm
Copper (Cu) Max. 0.002 %
Iron (Fe) Max. 10 ppm
Mercury (Hg) Max. 1 ppm
Lead (Pb) Max. 2 ppm
Zinc (Zn) Max. 0.002 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 100 ppm
Sulfate (SO₄) Max. 50 ppm
Alkali hydroxide, -bicarbonates complying
Appearance of the solution complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR