n-Hexane 99, reagent

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

139386-100ML 139386-2.5L 139386-500ML 139386-6X500ML
HONC139386-100MLEA 214 SGD
HONC139386-100ML HONC139386-2.5L HONC139386-500ML HONC139386-6X500ML
n-Hexane 99, reagent
n-Hexane

  • ≥95% for general laboratory use.


Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 66…69 °C(1.013 hPa)
Melting Pt: –95 °C
Density: 0.660…0.668 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.0 %
Refractive index (n 20/D) 1.373 - 1.377
Vapor Pressure ~132 mmHg ( 20 °C)
Vapor Density ~3 (vs air)
Refractive Index n20/D 1.375 (lit.)
Explosion Limit 7.7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR