Formic acid ~98% (by titrimetric analysis), LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Aminic Acid, Methanoic acid

14265-1ML
HONC14265-1MLEA 287 SGD
HONC14265-1ML
Formic acid ~98% (by titrimetric analysis), LC-MS Ultra eluent additive, tested for UHPLC-MS, Fluka™
Formic acid
Formula: HCO₂H
MW: 46.03 g/mol
Boiling Pt: 100.4 °C (1.013 hPa)
Melting Pt: 4 °C
Density: 1.22 g/cm3 (20 °C)
Flash Pt: 49.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1779
ADR: 8 II
REACH: 01-2119491174-37
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 97.5 %
Suitability for the LC-MS complying
Non-volatile matter Max. 20 ppm
Silver (Ag) Max. 0.02 ppm
Aluminium (Al) Max. 0.05 ppm
Barium (Ba) Max. 0.05 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.2 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.05 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.2 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.02 ppm
Magnesium (Mg) Max. 0.5 ppm
Manganese (Mn) Max. 0.05 ppm
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.05 ppm
Lead (Pb) Max. 0.02 ppm
Strontium (Sr) Max. 0.02 ppm
Thallium (Tl) Max. 0.05 ppm
Zinc (Zn) Max. 0.05 ppm
Chloride (Cl) Max. 5 ppm
Sulfate (SO₄) Max. 5 ppm
Sulfite (SO₃) Max. 10 ppm
Ammonium (NH₄) Max. 10 ppm
APHA Max. 15
Appearance of the solution complying
Autoignition Temperature 1004 °F

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR