Hexane (mixture of isomers) ≥85% (isomers basis, GC), Puriss.

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexanes, Isohexane

15613-2.5L 15613-25L
HONC15613-2.5LEA 177 SGD
HONC15613-2.5L HONC15613-25L
Hexane (mixture of isomers) ≥85% (isomers basis, GC), Puriss.
Hexane (mixture of isomers)

Beilstein Number: 1730733

Formula: H₃C(CH₂)₄CH₃
Boiling Pt: 48…65 °C (1,013 hPa)
Melting Pt: –154 °C
Density: 0,66 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c38497992e162315d0f3563bfcd3dc478422095&vid=7880d919bf16a88cca44241139a2be8054e77804&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay of isomers (GC) Min. 85 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.002 %
Non-volatile matter Max. 0.002 %
Aromatic substances (as C₆H₆) (GC) Max. 0.01 %
S-compounds (as S) Max. 0.005 %
n-Hexane (GC) Max. 4.9 %
Autoignition Temperature 437 °F
Vapor Density 3 (vs air)
Explosion Limit 7.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR