Ethyl acetate ≥99.0% (by GC), Laboratory Reagent

Supplier: Honeywell Chemicals
Danger

Synonyms: Acetic acid ethyl ester

16371-4X5L
HONC16371-4X5LEA 2770 SGD
HONC16371-4X5L
Ethyl acetate ≥99.0% (by GC), Laboratory Reagent
Ethyl acetate

  • ≥95% for general laboratory use.


Beilstein Number: 506104
RTECS Number: AH5425000
UNSPSC Code: 12191602

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77 °C (1.013 hPa)
Melting Pt: –84 °C
Density: 0.901 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.0 %
Free acid (as CH₃COOH) Max. 0.003 %
Water (Karl Fischer) Max. 0.05 %
Non-volatile matter Max. 0.003 %
Explosion Limit 2.2-11.5 %, 38 °F
Refractive Index n20/D 1.3720 (lit.)
Autoignition Temperature 801 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR