Magnesium sulphate, anhydrous ≥97%, Reagent Grade

Supplier: Honeywell Chemicals

208094-2.5KG
HONC208094-2.5KGEA 1130 SGD
HONC208094-2.5KG
Magnesium sulphate, anhydrous ≥97%, Reagent Grade
Magnesium sulphate

  • ≥95% for general laboratory use.

Formula: MgSO₄
Melting Pt: 1,130 °C (dec.)
Density: 2,7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay of Mg 19.6 - 20.8 %
Vapor Pressure <0.1 mmHg ( 20 °C)
Vapor Density <0.01 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR