Hexane (mixture of isomers) 95, Laboratory Reagent

Supplier: Honeywell Chemicals
Danger

Synonyms: Hexanes, Isohexane

208752-2.5L 208752-25L 208752-1L 208752-4X2.5L 208752-6X1L
HONC208752-2.5LEA 613 SGD
HONC208752-2.5L HONC208752-25L HONC208752-1L HONC208752-4X2.5L HONC208752-6X1L
Hexane (mixture of isomers) 95, Laboratory Reagent
Hexane (mixture of isomers)

  • ≥95% for general laboratory use.


Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 66…69 °C(1.013 hPa)
Melting Pt: –95 °C
Density: 0.660…0.668 g/cm³ (20 °C)
Flash Pt: –26 °C
Storage Temperature: Ambient
MDL Number: MFCD02179311
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Autoignition Temperature 453 °F
IR spectrum complying
Assay (GC) Min. 95.0 %
Density (D 20/20) 0.658 - 0.662
Refractive index (n 20/D) 1.373 - 1.377
Non-volatile matter Max. 0.0015 %
Water (Karl Fischer) Max. 0.015 %
Aromatic substances (as C₆H₆) Max. 0.005 %
Vapor Pressure 5.2 psi ( 37.7 °C)
Explosion Limit 7.7 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR