Potassium permanganate ≥99.0% ACS, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Permanganic acid potassium salt

223468-25G 223468-500G
HONC223468-25GEA 210 SGD
HONC223468-25G HONC223468-500G
Potassium permanganate ≥99.0% ACS, Fluka™
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Melting Pt: Dec.
Density: 2.700 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1490
ADR: 5.1 II
Merck Index: 12,07824

Order Now

Specification Test Results

Assay Min. 99.0 %
Water insoluble matter Max. 0.2 %
Chloride, Chlorate (as Cl) Max. 0.005 %
Sulfate (SO₄) Max. 0.02 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR