Lead(II) nitrate ≥99.0% ACS, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Lead dinitrate

228621-100G 228621-2.5KG 228621-500G
HONC228621-100GEA 158 SGD
HONC228621-100G HONC228621-2.5KG HONC228621-500G
Lead(II) nitrate ≥99.0% ACS, Fluka™
Lead (II) nitrate
Formula: Pb(NO₃)₂
MW: 331.21 g/mol
Boiling Pt: Dec. (on heating)
Melting Pt: 458…459 °C (1.023 hPa)
Density: 4.49 g/cm³ (20.1 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1 II
Merck Index: 13,05432

Order Now

Specification Test Results

Assay 99.0 - 101.0 %
In diluted HNO₃ insoluble matter Max. 0.005 %
Calcium (Ca) Max. 50 ppm
Copper (Cu) Max. 20 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 50 ppm
Sodium (Na) Max. 200 ppm
Chloride (Cl) Max. 10 ppm
Form Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR