Cobalt(II) chloride hexahydrate 98% ACS, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

255599-100G 255599-500G 255599-5G
HONC255599-100GEA 298 SGD
HONC255599-100G HONC255599-500G HONC255599-5G
Cobalt(II) chloride hexahydrate 98% ACS, Fluka™
Cobalt (II) chloride hexahydrate
Formula: CoCl₂·6H₂O
Boiling Pt: Decomposes on heating,
Melting Pt: 86 °C
Density: 1,9 g/cm³ (20 °C)
Storage Temperature: Room Temperature
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
ADR: 9,III
Merck Index: 13,02462

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 0.002 %
Iron (Fe) Max. 0.005 %
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.005 %
Sodium (Na) Max. 0.05 %
Nickel (Ni) Max. 0.1 %
Zinc (Zn) Max. 0.03 %
Nitrate (NO₃) Max. 0.01 %
Sulfate (SO₄) Max. 0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR