Water, CHROMASOLV™ for HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals

Synonyms: Hydrogen oxide

270733-1L 270733-2.5L 270733-4L 270733-6X1L
HONC270733-1LEA 173 SGD
HONC270733-1L HONC270733-2.5L HONC270733-4L HONC270733-6X1L
Water, CHROMASOLV™ for HPLC, Riedel-de Haën™
Water

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.9971 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Non-volatile matter Max. 0.0003 %
Absorbance at 400 nm Max. 0.005
Absorbance at 250 nm Max. 0.005
Absorbance at 210 nm Max. 0.01
Absorbance at 205 nm Max. 0.01
HPLC Gradient at 254 nm Max. 2 mAU
HPLC Gradient at 210 nm Max. 10 mAU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR