ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: Reagent alcohol

270741-1L
HONC270741-1LEA 816 SGD
HONC270741-1L
ALCH, CHROMASOLV™ for HPLC (denatured with 5% isopropanol and 5% methanol), Riedel-de Haën™
Alcohol

Reagent Alcohol contains 95 parts of specially denatured ethyl alcohol (SDA) 3A, with 5 parts of isopropyl alcohol. Final concentrations are ~ 90% ethanol, ~ 5% methanol and ~ 5% isopropanol.


  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency

Boiling Pt: 78,32 °C (main component)
Melting Pt: –114,1 °C (main component)
Density: 0,7890 g/cm³ (20 °C)
Flash Pt: 15 °C (main component)
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 89.0 %
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.1 %
Methanol (GC) 3.0 - 5.0 %
2-propanol (GC) 4.0 - 6.0 %
Absorbance at 400 nm Max. 0.01
Absorbance at 300 nm Max. 0.01
Absorbance at 255 nm Max. 0.04
Absorbance at 220 nm Max. 0.35
Absorbance at 210 nm Max. 0.65
Absorbance at 205 nm Max. 1.00


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR